siRNA Details
Gene Symbol: Ppat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172146.2 | NP_742158.1 | 4, 5 | 1013 | ||
GenBank mRNA | AK147750.1 | - | 4, 5 | 663 | ||
AK148034.1 | - | 4, 5 | 617 | |||
AK159141.1 | - | 5, 6 | 1163 | |||
AK159859.1 | - | 5, 6 | 1163 | |||
AK160542.1 | - | 4, 5 | 638 | |||
AK164506.1 | - | 4, 5 | 627 | |||
AK167057.1 | - | 4, 5 | 652 | |||
BC006053.1 | - | 4, 5 | 600 | |||
BC023841.1 | - | 4, 5 | 640 | 5346105 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
