siRNA Details
Gene Symbol: Plac8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139198.2 | NP_631937.1 | 3 | 304 | ||
GenBank mRNA | AF263458.1 | - | 3 | 292 | ||
AJ422151.1 | - | 3 | 324 | |||
AK007369.1 | - | 3 | 305 | |||
BC010789.1 | - | 3 | 275 | 4216402 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
