siRNA Details
Gene Symbol: Fbxo21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145564.3 | NP_663539.1 | 2 | 318 | ||
GenBank mRNA | AB093270.1 | - | 2 | 304 | ||
AK048131.1 | - | 2 | 297 | |||
AK048542.1 | - | 2 | 297 | |||
AK049723.1 | - | 2 | 319 | |||
BC021871.1 | - | 2 | 355 | 3709413 |

More Information
Other Species Targeted

Gene Ontology Categories:
