siRNA Details
Gene Symbol: Fam109a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175474.3 | NP_780683.1 | 4 | 1902 | ||
GenBank mRNA | AK033618.1 | - | 4 | 1903 | ||
AK039192.1 | - | 3 | 1850 | |||
AK081460.1 | - | 1 | 1023 | |||
BC095929.1 | - | 327 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
