siRNA Details
Gene Symbol: B3gnt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198611.2 | NP_941013.2 | 1 | 422 | ||
GenBank mRNA | AY037786.1 | - | 1 | 423 | ||
BC114987.1 | - | 1 | 377 | 40056240 | ||
BC115755.1 | - | 1 | 377 | 40056241 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
