siRNA Details
Gene Symbol: Agfg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK155221.1 | - | 3 | 447 | ||
AK171048.1 | - | 3 | 492 | |||
BC003330.1 | - | 3 | 459 | 3256586 | ||
BC046788.1 | - | 3 | 383 | 5716604 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
