siRNA Details
Gene Symbol: Tmem184a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161548.1 | NP_001155020.1 | 7 | 1000 | ||
NM_144914.3 | NP_659163.3 | 6 | 820 | |||
GenBank mRNA | AK050489.1 | - | 7 | 970 | ||
AK148454.1 | - | 792 | ||||
BC019731.1 | - | 2 | 209 | 3993397 | ||
BC026659.1 | - | 7 | 959 | 4988857 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
