siRNA Details
Gene Symbol: Aimp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172146.1 | NP_001165617.1 | 2 | 382 | ||
NM_146165.2 | NP_666277.1 | 2 | 262 | |||
GenBank mRNA | BC024410.1 | - | 2 | 355 | 4954972 | |
BC024480.1 | - | 2 | 253 | 4977193 | ||
BC026958.1 | - | 2 | 321 | 4013019 | ||
BC026972.1 | - | 2 | 335 | 5371230 | ||
CF732613.1 | - | 2 | 304 | |||
CJ051555.1 | - | 2 | 254 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
