siRNA Details
Gene Symbol: Plekha8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001335.2 | NP_001001335.1 | 7 | 905 | ||
NM_001164361.1 | NP_001157833.1 | 7 | 1000 | |||
GenBank mRNA | AK078040.1 | - | 7 | 1001 | ||
AK170592.1 | - | 7 | 767 | |||
BC052360.1 | - | 7 | 905 | 6704789 |
