siRNA Details
Gene Symbol: Vopp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146168.1 | NP_666280.1 | 2 | 234 | ||
GenBank mRNA | AK030975.1 | - | 2 | 221 | ||
AK031812.1 | - | 2 | 239 | |||
AK158041.1 | - | 246 | ||||
AK165256.1 | - | 2 | 263 | |||
AK170152.1 | - | 2 | 185 | |||
AK170832.1 | - | 2 | 185 | |||
BC024822.1 | - | 2 | 234 | 5359195 | ||
BC024884.1 | - | 2 | 156 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
