siRNA Details
Gene Symbol: Eva1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145570.2 | NP_663545.2 | 3 | 1341 | ||
GenBank mRNA | AK145140.1 | - | 3 | 1350 | ||
BC014699.1 | - | 3 | 1275 | 4221746 | ||
BC027150.1 | - | 3 | 1297 | 4950341 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
