siRNA Details
Gene Symbol: Eva1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145570.2 | NP_663545.2 | 2 | 451 | ||
GenBank mRNA | AK145140.1 | - | 2 | 457 | ||
BC014699.1 | - | 2 | 384 | 4221746 | ||
BC027150.1 | - | 2 | 406 | 4950341 | ||
CK020834.1 | - | 2 | 372 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
