siRNA Details
Gene Symbol: Mob1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145571.2 | NP_663546.1 | 5, 6 | 791 | ||
GenBank mRNA | AK028416.1 | - | 5, 6 | 814 | ||
AK028695.1 | - | 3, 4 | 524 | |||
AK158228.1 | - | 5, 6 | 775 | |||
AK167515.1 | - | 5, 6 | 786 | |||
AK170691.1 | - | 5, 6 | 786 | |||
AK172205.1 | - | 5, 6 | 794 | |||
BC009149.1 | - | 5, 6 | 791 | 2651599 | ||
BC033463.1 | - | 5, 6 | 745 | 3500664 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001033891, BC101879.
Gene Ontology Categories:
