siRNA Details
Gene Symbol: Vgll4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177683.2 | NP_808351.1 | 5 | 1270 | ||
GenBank mRNA | AK144447.1 | - | 5 | 1313 | ||
AK162019.1 | - | 4 | 714 | |||
AK163550.1 | - | 6 | 1254 | |||
AK181016.1 | - | 1 | 138 | |||
BC048841.1 | - | 5 | 1270 | 5008915 | ||
BC060305.1 | - | 5 | 1283 | 3586396 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
