siRNA Details
Gene Symbol: A2m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175628.3 | NP_783327.2 | 26 | 3285 | ||
GenBank mRNA | AY185125.1 | - | 26 | 3231 | ||
BC023844.1 | - | 16 | 1947 | |||
BC072642.1 | - | 26 | 3225 | 6839761 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
