siRNA Details
Gene Symbol: A2m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175628.3 | NP_783327.2 | 10 | 1149 | ||
GenBank mRNA | AY185125.1 | - | 10 | 1098 | ||
BC072642.1 | - | 10 | 1089 | 6839761 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
