siRNA Details
Gene Symbol: A2m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175628.3 | NP_783327.2 | 28 | 3546 | ||
GenBank mRNA | AY185125.1 | - | 28 | 3492 | ||
BC023844.1 | - | 18 | 2208 | |||
BC072642.1 | - | 28 | 3486 | 6839761 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
