siRNA Details
Gene Symbol: BC048546 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001179.3 | NP_001001179.2 | 17 | 2115 | ||
GenBank mRNA | AK138836.1 | - | 17 | 2116 | ||
AK161068.1 | - | 4 | 465 | |||
AK161184.1 | - | 17 | 2118 | |||
BC040781.1 | - | 5 | 567 |
