siRNA Details
Gene Symbol: BC048546 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001179.3 | NP_001001179.2 | 31 | 4078 | ||
GenBank mRNA | AK138836.1 | - | 31 | 4079 | ||
AK161068.1 | - | 18 | 2428 | |||
BC034132.1 | - | 6 | 838 | |||
BC040781.1 | - | 19 | 2530 |
