siRNA Details
Gene Symbol: Klhl42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081237.1 | NP_001074706.1 | 2, 3 | 1490 | ||
GenBank mRNA | AK048908.1 | - | 2, 3 | 1079 | ||
AK052765.1 | - | 2, 3 | 1067 | |||
AK083047.1 | - | 2, 3 | 1075 | |||
AK129335.1 | - | 2, 3 | 400 | |||
BC037084.1 | - | 2, 3 | 1095 | |||
BC147633.1 | - | 2, 3 | 1246 | 9056623 | ||
BC147636.1 | - | 2, 3 | 1245 | 9056626 | ||
BC151064.1 | - | 2, 3 | 1246 | 9087977 | ||
BC151065.1 | - | 2, 3 | 1246 | 9087978 | ||
BC151068.1 | - | 2, 3 | 1246 | 9087981 | ||
BC151076.1 | - | 2, 3 | 1246 | 9087989 |
