siRNA Details
Gene Symbol: Amn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113424.1 | NP_001106895.1 | 4, 5 | 601 | ||
GenBank mRNA | AK038995.1 | - | 2, 3 | 647 | ||
AK050169.1 | - | 4, 5 | 942 | |||
AK053764.1 | - | 1, 2 | 2536 | |||
BC026924.1 | - | 2, 3 | 284 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
