siRNA Details
Gene Symbol: Amn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113424.1 | NP_001106895.1 | 5, 6 | 660 | ||
GenBank mRNA | AK038995.1 | - | 3, 4 | 706 | ||
AK050169.1 | - | 5, 6 | 1001 | |||
AK053764.1 | - | 2, 3 | 2595 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
