siRNA Details
Gene Symbol: Gm4876 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK047082.1 | - | 1 | 317 | ||
BC120768.1 | - | 1 | 293 | 40129691 | ||
BC120770.1 | - | 1 | 296 | 40129693 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
