siRNA Details
Gene Symbol: Mgam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171003.1 | NP_001164474.1 | 47 | 5533 | ||
GenBank mRNA | AK031332.1 | - | 31 | 3536 | ||
AK033548.1 | - | 7 | 672 | |||
BC025509.1 | - | 13 | 1233 | |||
EU937529.1 | - | 47 | 5533 |
