siRNA Details
Gene Symbol: Leng8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172736.3 | NP_766324.1 | 4 | 435 | ||
GenBank mRNA | AK034310.1 | - | 4 | 440 | ||
AK085304.1 | - | 4 | 433 | |||
AK170629.1 | - | 4 | 443 | |||
AK200008.1 | - | 1 | 72 | |||
BC041775.1 | - | 4 | 407 | |||
BC042658.1 | - | 4 | 423 | 5365041 | ||
BC066768.1 | - | 4 | 426 | 6407146 | ||
CF536188.1 | - | 4 | 435 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
