siRNA Details
Gene Symbol: Suv420h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115018.1 | NP_001108490.1 | 9 | 1444 | ||
NM_146177.2 | NP_666289.2 | 9 | 1502 | |||
GenBank mRNA | AK137449.1 | - | 9 | 1448 | ||
AK154848.1 | - | 9 | 2042 | |||
AY555193.1 | - | 9 | 1456 | |||
BC024816.1 | - | 9 | 1441 | 5359433 | ||
BC085473.1 | - | 7 | 1198 | 6414100 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
