siRNA Details
Gene Symbol: Shisa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290291.1 | NP_001277220.1 | 3 | 1046 | ||
NM_172737.4 | NP_766325.3 | 3 | 1046 | |||
GenBank mRNA | AK085127.1 | - | 2 | 268 | ||
AK145016.1 | - | 2 | 331 | |||
BY734785.1 | - | 2 | 331 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
