siRNA Details
Gene Symbol: PMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000534.4 | NP_000525.1 | 10 | 2421 | ||
NM_001128143.1 | NP_001121615.1 | 9 | 2304 | |||
NM_001289408.1 | NP_001276337.1 | 9 | 2318 | |||
NM_001289409.1 | NP_001276338.1 | 8 | 2135 | |||
NM_001321045.1 | NP_001307974.1 | 11 | 2547 | |||
NM_001321046.1 | NP_001307975.1 | 9 | 2238 | |||
NM_001321047.1 | NP_001307976.1 | 10 | 2598 | |||
NM_001321048.1 | NP_001307977.1 | 11 | 2518 | |||
NR_110332.1 | 10 | 2401 | ||||
XM_006712596.1 | XP_006712659.1 | 6 | 1386 | |||
XM_011511356.2 | XP_011509658.1 | 10 | 2707 | |||
XM_017004344.1 | XP_016859833.1 | 10 | 2030 | |||
XM_017004345.1 | XP_016859834.1 | 9 | 1855 | |||
XM_017004346.1 | XP_016859835.1 | 9 | 1818 | |||
XM_017004347.1 | XP_016859836.1 | 8 | 1721 | |||
XM_017004349.1 | XP_016859838.1 | 9 | 2590 | |||
GenBank mRNA | AB102869.1 | - | 8 | 1775 | ||
AB102875.1 | - | 8 | 1775 | |||
AK295602.1 | - | 9 | 1914 | |||
AK297440.1 | - | 10 | 1949 | |||
AK304634.1 | - | 9 | 1757 | |||
AK316215.1 | - | 8 | 1655 | |||
AY540750.1 | - | 5 | 1410 | |||
AY540751.1 | - | 10 | 1973 | |||
BC036376.1 | - | 10 | 2140 | |||
BC096330.1 | - | 10 | 1962 | |||
BC096332.1 | - | 10 | 1962 | |||
BM553209.1 | - | 2 | 148 | |||
CR749432.1 | - | 1 | 120 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
