siRNA Details
Gene Symbol: Nlrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177690.3 | NP_808358.2 | 5 | 1820 | ||
GenBank mRNA | AK054264.1 | - | 5 | 1821 | ||
AY360473.1 | - | 5 | 1821 | |||
DQ066881.1 | - | 5 | 1821 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
