siRNA Details
Gene Symbol: Wdr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199060.1 | NP_001185989.1 | 9 | 1267 | ||
NM_001199061.1 | NP_001185990.1 | 9 | 999 | |||
NM_021312.5 | NP_067287.1 | 8 | 1089 | |||
GenBank mRNA | AB041608.1 | - | 9 | 1268 | ||
AF239765.1 | - | 8 | 745 | |||
AK007500.1 | - | 8 | 866 | |||
AK016611.1 | - | 9 | 943 | |||
AK031436.1 | - | 8 | 852 | |||
AK149085.1 | - | 8 | 771 | |||
AK161299.1 | - | 9 | 1012 | |||
AY059431.1 | - | 8 | 780 | |||
AY059432.1 | - | 1 | 85 | |||
BC004748.1 | - | 8 | 923 | 3583955 | ||
BC052386.1 | - | 8 | 1060 | 6418498 | ||
BC096651.1 | - | 8 | 834 | 1514703 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
