siRNA Details
Gene Symbol: Igfl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003393.1 | NP_001003393.1 | 3 | 392 | ||
GenBank mRNA | AY672115.1 | - | 3 | 392 | ||
BC147097.1 | - | 3 | 392 | 9056138 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
