siRNA Details
Gene Symbol: Mtmr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023858.3 | NP_076347.3 | 11 | 1507 | ||
GenBank mRNA | AK133476.1 | - | 11 | 1507 | ||
AK133889.1 | - | 10 | 1275 | |||
AK220369.1 | - | 4 | 7103 | |||
AY055832.1 | - | 11 | 2278 | |||
BC027013.1 | - | 2 | 1084 | |||
BC063050.1 | - | 12 | 1596 | 5716297 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
