siRNA Details
Gene Symbol: Ccdc61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033314.3 | NP_001028486.1 | 1 | 225 | ||
GenBank mRNA | AK082980.1 | - | 1 | 220 | ||
AK146297.1 | - | 1 | 226 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
