siRNA Details
Gene Symbol: Zfp114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029933.2 | NP_001025104.2 | 4 | 873 | ||
GenBank mRNA | AF167317.1 | - | 4 | 858 | ||
AK139475.1 | - | 3 | 1792 | |||
AY166793.1 | - | 2 | 635 | |||
BC096771.1 | - | 2 | 1099 | |||
BC138311.1 | - | 2 | 988 | 8861331 | ||
BC138312.1 | - | 2 | 988 | 8861332 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
