siRNA Details
Gene Symbol: Phldb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102613.1 | NP_001096083.1 | 6 | 952 | ||
GenBank mRNA | BC052555.1 | - | 1 | 256 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Phldb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102613.1 | NP_001096083.1 | 6 | 952 | ||
GenBank mRNA | BC052555.1 | - | 1 | 256 |