siRNA Details
Gene Symbol: Lypd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182785.3 | NP_877586.1 | 5 | 900 | ||
GenBank mRNA | AK077071.1 | - | 5 | 901 | ||
BC049744.1 | - | 4 | 700 | 6772266 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
