siRNA Details
Gene Symbol: B3gnt8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001036740.2 | NP_001031817.1 | 2 | 767 | ||
NM_146184.4 | NP_666296.1 | 1 | 1159 | |||
GenBank mRNA | AK042388.1 | - | 1 | 1142 | ||
AK154220.1 | - | 754 | ||||
AK154901.1 | - | 2 | 723 | |||
BC025206.1 | - | 2 | 753 | 5037295 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
