siRNA Details
Gene Symbol: B9d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172148.1 | NP_742160.1 | 2 | 423 | ||
GenBank mRNA | AK141500.1 | - | 3 | 398 | ||
AK146227.1 | - | 3 | 371 | |||
BC028440.1 | - | 2 | 423 | 1265537 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
