siRNA Details
Gene Symbol: Cyp2a22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101467.1 | NP_001094937.1 | 6 | 946 | ||
GenBank mRNA | AK149509.1 | - | 5 | 2166 | ||
BC151116.1 | - | 6 | 940 | 9088029 | ||
BC158034.1 | - | 6 | 940 | 9007337 |
