siRNA Details
Gene Symbol: Hipk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033315.2 | NP_001028487.2 | 2 | 1056 | ||
GenBank mRNA | AB354583.1 | - | 2 | 800 | ||
AK133498.1 | - | 1 | 1573 | |||
BC147572.1 | - | 1 | 216 | 9056562 | ||
BC147577.1 | - | 1 | 216 | 9056567 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
