siRNA Details
Gene Symbol: Fbxo27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163702.1 | NP_001157174.1 | 4 | 826 | ||
NM_207238.3 | NP_997121.1 | 5 | 821 | |||
GenBank mRNA | AK053292.1 | - | 5 | 823 | ||
AK140275.1 | - | 4 | 824 | |||
AK158463.1 | - | 4 | 827 | |||
AK169614.1 | - | 4 | 911 | |||
BC075656.1 | - | 4 | 789 | 30540609 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
