siRNA Details
Gene Symbol: Rasgrp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174155.1 | NP_001167626.1 | 6 | 719 | ||
NM_145149.4 | NP_660131.2 | 6 | 719 | |||
GenBank mRNA | AF331457.1 | - | 6 | 719 | ||
AK049562.1 | - | 5 | 462 | |||
AK087348.1 | - | 6 | 672 | |||
AK089282.1 | - | 6 | 666 | |||
AK154746.1 | - | 6 | 709 | |||
AK162667.1 | - | 6 | 720 | |||
AK165106.1 | - | 5 | 457 | |||
AY040628.1 | - | 6 | 719 | |||
AY196476.1 | - | 6 | 579 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
