siRNA Details
Gene Symbol: Zfp940 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173738.2 | NP_776099.1 | 5 | 1673 | ||
GenBank mRNA | AK132317.1 | - | 4 | 1912 | ||
AK139326.1 | - | 4 | 1936 | |||
AK141787.1 | - | 1966 | ||||
BC027344.1 | - | 5 | 1673 | 3709361 |
