siRNA Details
Gene Symbol: Zfp420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172740.2 | NP_766328.2 | 4 | 1687 | ||
GenBank mRNA | AB010367.1 | - | 1 | 218 | ||
AK045435.1 | - | 2 | 1345 | |||
AK045821.1 | - | 4 | 1600 | |||
BC050249.1 | - | 3 | 1572 | |||
BC055817.1 | - | 4 | 1607 | 3595825 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
