siRNA Details
Gene Symbol: U2af1l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170760.3 | NP_739566.1 | 2 | 123 | ||
GenBank mRNA | AF419339.1 | - | 2 | 107 | ||
BC051477.1 | - | 2 | 131 | |||
BC060972.1 | - | 2 | 123 | 6593404 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
