siRNA Details
Gene Symbol: 4931406P16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172741.2 | NP_766329.1 | 9 | 2305 | ||
GenBank mRNA | AK076994.1 | - | 9 | 2306 | ||
AK129125.1 | - | 8 | 1283 | |||
AK170198.1 | - | 10 | 1521 | |||
BC060233.1 | - | 9 | 2106 | 6852905 |

More Information
Other Species Targeted

Gene Ontology Categories:
