siRNA Details
Gene Symbol: Lim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177693.3 | NP_808361.1 | 4, 5 | 519 | ||
GenBank mRNA | AK029354.1 | - | 4, 5 | 520 | ||
AK164958.1 | - | 4, 5 | 518 | |||
BC092386.1 | - | 4, 5 | 532 | 30551056 | ||
BC119253.1 | - | 4, 5 | 500 | 8734002 | ||
BC119255.1 | - | 4, 5 | 500 | 8734004 |

More Information
Other Species Targeted

Gene Ontology Categories:
