siRNA Details
Gene Symbol: Prr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175022.2 | NP_778187.2 | 4 | 856 | ||
GenBank mRNA | AK173130.1 | - | 1 | 295 |

More Information
Other Species Targeted

Gene Ontology Categories:
