siRNA Details
Gene Symbol: Mrgpra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153067.2 | NP_694707.2 | 2 | 657 | ||
GenBank mRNA | AY042193.1 | - | 2 | 672 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mrgpra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153067.2 | NP_694707.2 | 2 | 657 | ||
GenBank mRNA | AY042193.1 | - | 2 | 672 |