siRNA Details
Gene Symbol: Synm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ579700.1 | - | 4 | 4435 | ||
AJ579701.1 | - | 4, 5 | 3529 | |||
AJ579702.1 | - | 3, 4 | 1082 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
